Copper metal ≥99%, fine powder

Warning

C1335-2.5KG C1335-500GM C1335-125GM
700013-290EA 220.9 USD
700013-290 700013-284 700013-286
Copper metal ≥99%, fine powder
Copper

Copper Metal, Fine Powder is an element with the symbol Cu and atomic number 29. It is a ductile metal with very high thermal and electrical conductivity. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2567 °C (1013 hPa)
Melting Pt: 1083.4 °C
Density: 8,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR