Calcium carbonate ≥98%, heavy powder FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Marble
Calcium Carbonate, Heavy Powder, FCC is used as an acidity regulator, anti-caking agent, preservative and stabilizer in some applications like flour and enriched rice. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
CaCO₃ MW: 100.09 g/mol Melting Pt: 825 °C Density: 2.71 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...