Isobutyl acetate ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: 2-Methylpropyl acetate

B2321-500MLG
700004-430EA 109 USD
700004-430
Isobutyl acetate ≥98%
Isobutyl acetate

iso-Butyl Acetate has a combination of good solvent activity, desirable evaporation rate, and pleasant odor enables it to be used in a wide range of coating applications. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.868
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008932
CAS Number: 110-19-0
EINECS: 203-745-1
UN: 1213
ADR: 3,II
Merck Index: 13,05147

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR