2,3-Butanediol

Synonyms: 2,3-Dihydroxy butane2,3-Butylene glykol(±)-2,3-Butanediol

B1590-25ML B1590-500ML B1590-100ML
700002-924EA 571.74 USD
700002-924 700002-922 700002-920
2,3-Butanediol
2,3-Butanediol

2,3-Butanediol, also known as Dimethylene glycol, is used in the resolution of carbonyl compounds in gas chromatography.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 184 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 0.994
Storage Temperature: Ambient
MDL Number: MFCD00004523
CAS Number: 513-85-9
EINECS: 208-173-6
Merck Index: 12,01602

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR