Butyl stearate

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Stearic acid butyl esterButyl octadecanoate

B1192-500MLG B1192-200LTB B1192-100ML B1192-4LTGL B1192-20LTMT
700004-968EA 68.12 USD
700004-968 700004-976 700004-970 700004-974 700004-972
Butyl stearate
Butyl stearate

Butyl Stearate is used as a solvent, spreading and softening agent in plastics, textiles, cosmetics, rubber industries.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₂H₄₄O₂
MW: 340.59 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 21 °C
Density: 0.854 g/cm³ (20 °C)
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00026669
CAS Number: 123-95-5
EINECS: 204-666-5
Merck Index: 12,01625

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR