2-Butoxyethyl acetate

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Butylglycol acetate

B1181-1LTGL B1181-20LTMT B1181-4LTGL
700004-418EA 1491 USD
700004-418 700004-416 700004-420
2-Butoxyethyl acetate
2-Butoxyethyl acetate

2-Butoxyethyl Acetate is a solvent that is typically used in pharmaceuticals, sunscreens, cosmetics, inks, dyes and water based paints.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.942
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00009457
CAS Number: 112-07-2
EINECS: 203-933-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR