AMP (2-Amino-2-methylpropanol)
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
2-Amino-2-methyl-1-propanol
, AMP
AM156-20LTMT
AM156-200LTB
AM156-100ML
700006-994EA
86.5
USD
700006-994
700007-000
700006-998
AMP (2-Amino-2-methylpropanol)
AMP (2-Amino-2-methylpropanol)
2-Amino-2-methyl-1-propanol, or aminomethyl propanol is a colorless, viscous liquid that functions as a pH adjuster. It is also used as an intermediate in drug synthetic schemes.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
(CH₃)₂C(NH₂)CH₂OH MW: 89.14 g/mol Boiling Pt: 165 °C (1013 hPa) Melting Pt: 18…26 °C Density: 0.934 Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008051 CAS Number: 124-68-5 EINECS: 204-709-8 Merck Index: 13,00448 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...