4-Aminobutyric acid ≥98%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
γ-Aminobutyric acid
, GABA
4-Aminobutyric Acid, Reagent belongs to the transaminases family which transfer nitrogenous groups. It is an enzyme which plays a part in 5 metabolic pathways: glutamate metabolism, beta-alanine metabolism, alanine and aspartate metabolism, butanoate metabolism, and propionate metabolism. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 248 °C (1013 hPa) Melting Pt: 202 °C Density: 1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008226 CAS Number: 56-12-2 EINECS: 200-258-6 Merck Index: 13,00429 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...