Aluminium ammonium sulfate dodecahydrate ≥99.5%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Aluminium ammonium bis(sulphate) dodecahydrateAluminium ammonium bis(sulfate) dodecahydrate

AL224-100LBB AL224-12KG AL224-2.5KG
95031-716EA 658.64 USD
95031-716 700001-580 700001-582
Aluminium ammonium sulfate dodecahydrate ≥99.5%, powder FCC
Aluminium ammonium sulphate dodecahydrate

Aluminum Ammonium Sulfate, Dodecahydrate, Powder, FCC is used in foods as a buffer and neutralizing agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: AlNH₄(SO₄)₂·12H₂O
MW: 453.33 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 93 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149958
CAS Number: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325

Order Now

Specification Test Results

Assay 99.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR