Alizarine

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 1,2-Dihydroxyanthraquinone1,2-Dihydroxy-9,10-anthracenedione1,2-Dihydroxy-9,10-anthraquinone

AL185-25GM AL185-500GM AL185-100GM
700004-164EA 125.08 USD
700004-164 700004-168 700004-166
Alizarine
Alizarine

Alizarin, also known as 1,2-dihydroxyanthraquinone, can be used as a dye for textile fabrics.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Boiling Pt: 430 °C (1013 hPa)
Melting Pt: 290 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001201
CAS Number: 72-48-0
EINECS: 200-782-5
Merck Index: 13,00246

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR