Arachidyl alcohol ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Eicosanol

A3850-1GM A3850-5GM A3850-5KGBL A3850-25GM
700002-736EA 21877 USD
700002-736 700002-734 700002-732 700002-730
Arachidyl alcohol ≥95%
Arachidyl alcohol

Arachidyl Alcohol, also known as Arachidalcohol or 1-icosanol, is a straight-chain fatty alcohol used as an emollient in cosmetics.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: H₃C(CH₂)₁₈CH₂OH
MW: 298.55 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 71.5 °C
Density: 0.88 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002938
CAS Number: 629-96-9
EINECS: 211-119-4

Order Now

Specification Test Results

Assay 95% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR