Patent blue V calcium salt

Synonyms: Bis[hydrogen [4-[4-(diethylamino)-5'-hydroxy-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium], calcium saltBis[hydrogen [4-[4-(diethylamino)-5'-hydroxy-2',4'-disulfonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium], calcium saltAcid Blue 3 Calcium Salt

A3001-100GM A3001-25GM
700009-976EA 963.56 USD
700009-976 700009-978
Patent blue V calcium salt
Patent blue V calcium salt

Acid Blue 3 Calcium Salt is a biological stain double labeled product for electron microscopy and fluorescence research.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅₄H₆₂CaN₄O₁₄S₄
MW: 1159.45 g/mol
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00038746
CAS Number: 3536-49-0
EINECS: 222-573-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR