3-Aminophenol
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
m-Aminophenol
, 3-Hydroxyaniline
A2349-10KGBL
A2349-5GM
A2349-500GM
A2349-100GM
700007-124EA
126
USD
700007-124
700007-130
700007-126
700007-128
3-Aminophenol
3-Aminophenol
m-Aminophenol, also known as 3-Aminophenol or 3-Hydroxyaniline, is an organic compound and an aromatic amine which has applications as a stabilizer for chlorine-containing thermoplastics and as hair dye colorants.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 299 °C (1013 hPa) Melting Pt: 122 °C Density: 1.275 g/cm³ (20 °C) Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007786 CAS Number: 591-27-5 EINECS: 209-711-2 UN: 2512 ADR: 6.1,III Merck Index: 13,00459 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...