p-Anisic acid ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
4-Methoxybenzoic acid
, 4-Anisic acid
A2287-500GM
A2287-10KGBL
A2287-100GM
700006-610EA
3163
USD
700006-610
700006-608
700006-612
p-Anisic acid ≥98%
p-Anisic acid
p-Anisic Acid, also known as 4-methoxybenzoic acid or draconic acid, is used as an intermediate in the preparation of more complex organic compounds.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 275 °C (1013 hPa) Melting Pt: 182…185 °C Density: ∼1.385 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002542 CAS Number: 100-09-4 EINECS: 202-818-5 Merck Index: 13,00669 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...