Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

eVol® Digital Analytical Syringe, SGE

Supplier: Trajan Scientific
eVol®
2910205 2910200 2910100
eVol® is the coupling of two precision devices; a digitally controlled electronic drive and an XCHANGE® enabled analytical syringe

eVol® XR is also available in the eVol® NMR Edition. eVol® NMR Edition uses long needles that allows accurate manipulation of small volumes of sample enabling dilution directly in the analysis tube – even in tubes with the smallest internal diameters. eVol® NMR facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. These tubes can now be emptied for washing and re-use by using the eVol® unit.

It is designed to make everyone an expert because it standardizes liquid handling results independent of the operator’s skill. eVol® improves accuracy and reproducibility and speeds up as well as simplifies laboratory workflows. eVol® has a dispense range from 200 nL – 1mL using a single device and positive displacement analytical syringes enabling it to be used with the majority of solutions – organic, aqueous or hazardous. Because eVol® incorporates standard analytical syringes with needles it can pierce septa. Syringes are easily calibrated in-house or by an outside agency and calibration factors are stored in the electronic handle. Up to 20 methods and custom programs can be saved in the handle. eVol® is programmable to follow laboratory protocols and programs can be password protected. There are 4 functions; Aspirate/Dispense of a single volume, Repeat or Multiple Dispense, Manual Mode for titrations and Custom Programs to meet your laboratory workflows. Applications for eVol® include but are in no way limited to: Preparation of calibration standards, Preparation and addition of internal standards, Precise dispensing of aqueous and non-aqueous liquids, Routine dispensing, Ergonomic operation with substances in a fume hood, Sample dilution, Instrument injections, Eliminating the need for serial dilutions since smaller volumes can be accurately and precisely dispensed, Micro titration and TLC spotting.

Certifications: CE approved. Meets RoHS and WEE Directives.

Ordering information: Starter Kit includes eVol® XR Electronic Syringe Handle, Stand, Universal Charger, 3 eVol® Syringes - 5µL, 100µL, and 1mL, Comprehensive Manual, Quick Start Guide and CD with Manual in Multiple Languages. The eVol® NMR Edition includes 3 eVol® Syringes - 5µL with a 115mm needle, 50µL and 500µL syringes with both 115 and 180mm needles. Specifications are subject to change without notification.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
eVol® NMR Edition 2910100 89194-536 Each Retrieving Restricted
eVol ® XR, Handle only 2910205 89236-492 Each Retrieving Restricted
eVol® XR Syringe Starter Kit, 1.15 kg (40.7 oz.) 2910200 89236-490 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Accuracy 500 μL: ± 0.2 % for calibrated syringe at full scale, ± 0.5 % for calibrated syringe at 10% scale, ± 0.5 % for uncalibrated syringe at full scale; 100 μL: ± 0.2 % for calibrated syringe at full scale, ± 0.5 % for calibrated syringe at 10% scale, ± 0.7 % for uncalibrated syringe at full scale; 50 μL: ± 0.2 % for calibrated syringe at full scale, ± 0.5 % for calibrated syringe at 10% scale, ± 1.0 % for uncalibrated syringe at full scale; 5 μL: ± 0.2 % for calibrated syringe at full scale, ± 1.0 % for calibrated syringe at 10% scale, ± 1.0 % for uncalibrated syringe at full scale
Precision 500 μL: 0.3 % RSD at full scale, 0.6 % RSD at 10 % scale; 100 μL: 0.4 % RSD at full scale, 0.7 % RSD at 10 % scale; 50 μL: 0.4 % RSD at full scale, 0.8 % RSD at 10 % scale; 5 μL: 0.5 % RSD at full scale, 1.0 % RSD at 10 % scale

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
NM1/2.5(eVol® 500µL)-5 1/2.5ML NEEDLE PK5 (NDL: 23g, 50mm, Bevel Tip) 039110 14225-776 Pack of 5 Retrieving Restricted
NM1/2.5(eVol® 500µL)-LC 1/2.5ML NEEDLE PK5 (NDL: 22g, 51mm, LC Tip) 039250 14225-784 Pack of 5 Retrieving Restricted
N25/500-LC 500µL (eVol® 50µL) NEEDLE PK5 (NDL: 22g, 51mm, LC Tip) 038250 60362-236 Pack of 5 Retrieving Restricted
N25/500-5 25/500µL (eVol® 50µL) NEEDLE PK5 (NDL: 25g, 50mm, Bevel Tip) 38110 97049-438 Pack of 5 Retrieving Restricted
5µL eVol® SYRINGE (NDL: 25g, 50mm, Bevel Tip), 22 g (0.78 oz.) 2910020 97049-424 Each Retrieving Restricted
5µL eVol® SYRINGE 3 PACK (NDL: 25g, 50mm, Bevel Tip) 2910320 89205-480 Pack of 3 Retrieving Restricted
5µL eVol® SYRINGE NO NEEDLE, 22 g (0.78 oz.) 2910021 89205-470 Each Retrieving Restricted
50µL eVol® MEPS SYRINGE, 22 g (0.78 oz.) 2910027 89205-478 Each Retrieving Restricted
50µL eVol® SYRINGE (NDL: 25g, 50mm, Bevel Tip), 22 g (0.78 oz.) 2910022 97049-426 Each Retrieving Restricted
50µL eVol® SYRINGE 3 PACK (NDL: 25g, 50mm, Bevel Tip) 2910322 89205-482 Pack of 3 Retrieving Restricted
50µL eVol® SYRINGE NO NEEDLE, 22 g (0.78 oz.) 2910023 89205-472 Each Retrieving Restricted
500µL eVol® MEPS SYRINGE, 22 g (0.78 oz.) 2910026 89205-476 Each Retrieving Restricted
500µL eVol® SYRINGE (NDL: 23g, 50mm, Bevel Tip), 22 g (0.78 oz.) 2910024 97049-428 Each Retrieving Restricted
500µL eVol® SYRINGE 3 PACK (NDL: 23g, 50mm, Bevel Tip) 2910324 89205-484 Pack of 3 Retrieving Restricted
500µL eVol® SYRINGE NO NEEDLE, 22 g (0.78 oz.) 2910025 89205-474 Each Retrieving Restricted
6XSR XCHANGE SYRINGE RACK 031786 89194-552 Each Retrieving Restricted
eVol®Charger, 212 g (7.48 oz.) 2910012 97049-416 Each Retrieving Restricted
eVol®Charging Stand 2910030 97049-418 Each Retrieving Restricted
eVol® REPLACEMENT BATTERY 2910040 97049-420 Each Retrieving Restricted
eVol® STAND (PACKAGED), 88 g (3.10 oz.) 2910010 97049-422 Each Retrieving Restricted
eVol® Syringe with Bevel Tip, 100 µL, 25 gauge, 50 mm 2910029 89368-642 Each Retrieving Restricted
eVol® Syringe with Bevel Tip, 100 µL, 25 gauge, 50 mm 2910329 89368-640 Pack of 3 Retrieving Restricted
eVol® Syringe with Bevel Tip, 1 mL, 23 gauge, 50 mm, eVol® XR required 2910335 89236-494 Pack of 3 Retrieving Restricted
eVol® Syringe with MEPs Tip, 100 µL, 2011 Firmware Required 2910028 89368-644 Each Retrieving Restricted
eVol® XR 1 mL Syringe, 176 g (6.21 oz.) 2910035 89236-486 Each Retrieving Restricted
N25/500-11.5/0.63BV eVol® NEEDLE PK5 038161 14225-748 Pack of 5 Retrieving Restricted
N25/500-11.5C eVol® NEEDLE PK 2 038060 14225-738 Pack of 2 Retrieving Restricted
N25/500-5 25/500µL (eVol® 50µL) NEEDLE PK5 (NDL: 25g, 50mm, Bevel Tip) 038110 60361-324 Pack of 5 Retrieving Restricted
N25/500-7C 25/500µL (eVol® 50µL) NEEDLE PK2 (NDL: 25g, 70mm, Cone Tip) 038030 14225-736 Pack of 2 Retrieving Restricted
N25/500-NMR-18/0.63BV eVol® NEEDLE PK 2 038138 89194-540 Pack of 2 Retrieving Restricted
N25/500-NMR-18/0.63C eVol® NEEDLE PK 2 038038 89194-546 Pack of 2 Retrieving Restricted
N5-11.5/25BV 5µL eVol® NEEDLE PK2 036920 89194-538 Pack of 2 Retrieving Restricted
N5-11.5/25C 5µL eVol® NEEDLE PK2 036922 89194-544 Pack of 2 Retrieving Restricted
N5-5/23C 5µL eVol® NEEDLE PK5 (NDL: 23g, 50mm, Bevel Tip) 036914 89205-490 Pack of 5 Retrieving Restricted
N5-5/25BV 5µL eVol® NEEDLE PK5 (NDL: 25g, 50mm, Bevel Tip) 36910 97049-436 Pack of 5 Retrieving Restricted
N5-5/25BV 5µL eVol® NEEDLE PK5 (NDL: 25g, 50mm, Bevel Tip) 036910 89205-486 Pack of 5 Retrieving Restricted
N5-5.1/22LC 5µL eVol® NEEDLE PK5 036912 89205-488 Pack of 5 Retrieving Restricted
N5-7/25BV 5µL eVol® NEEDLE PK5 (NDL: 25g, 70mm, Bevel Tip) 036916 89205-492 Pack of 5 Retrieving Restricted
N5-7/26C 5µL eVol® NEEDLE PK5 (NDL: 26g, 70mm, Cone Tip) 036918 89205-494 Pack of 5 Retrieving Restricted
NM1/2.5(eVol®)-11.5 NEEDLE PK5 (NDL: 23g, 115mm, Bevel Tip) 039160 14225-782 Pack of 5 Retrieving Restricted
NM1/2.5(eVol® 500)-WLC 1/2.5ML NEEDLE PK5 (NDL: 25g, 50mm, LC Tip) 039260 14225-786 Pack of 5 Retrieving Restricted
NM1/2.5 (eVol® 500µL)-11.5/0.63C NEEDLE PK5 039060 89194-548 Pack of 5 Retrieving Restricted
NM1/2.5(eVol® 500µL)-5/0.63D NEEDLE PK2 (NDL: 23g, 50mm, Dome Tip) 039116 14225-778 Pack of 2 Retrieving Restricted
NM1/2.5(eVol® 500µL)-7 1/2.5ML NEEDLE PK5 (NDL: 23g, 70mm, Bevel Tip) 039130 14225-780 Pack of 2 Retrieving Restricted
NM1/2.5(eVol® 500µL)-NMR-18/0.63BV NEEDLE PK2 039138 89194-542 Pack of 2 Retrieving Restricted
NM1/2.5 (eVol® 500µL)-NMR-18/0.63C NEEDLE PK2 039038 89194-550 Pack of 2 Retrieving Restricted
P-500-eVol® 500µL PLUNGER 2910384 97049-434 Each Retrieving Restricted
P-50-eVol® 50µL PLUNGER 2910382 97049-432 Each Retrieving Restricted
P5-eVol® 5µL PLUNGER 2910380 97049-430 Each Retrieving Restricted
Replacement Needles with Side Holes, for 1-2.5 mL Removable Needle Syringes, 23 gauge, 50 mm 039120 10799-430 Pack of 2 Retrieving Restricted
Replacement Needle with Bevel Tip, 23 gauge, 56 mm, for 1 mL XCHANGE® GC Syringe 039125 10799-434 Pack of 5 Retrieving Restricted
Replacement Needle with LC Point Tip, 22 gauge, 56 mm, for 1 mL XCHANGE® LC Syringe 039256 10799-628 Pack of 5 Retrieving Restricted
Replacement Plunger, 100 µL, for 100 µL eVol® Syringe 2910383 89368-638 Each Retrieving Restricted
Replacement Plunger, 1 mL, for 1 mL eVol® Syringe 2910385 89236-496 Each Retrieving Restricted
Syringes with Bevel Tip, 1-2.5 mL, 22 gauge, 50 mm 039115 10799-432 Pack of 2 Retrieving Restricted
V25/2.5M-0.63C SYRINGE VALVE 031905 60361-256 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty