EcoPolyBlend™ Spill Control Accumulation Centers, Justrite
28652 28657 28660 28659 28655 28661 28654 28653 28656 28658
89107-290EA 409.81 USD
89107-290 89107-292 89107-294 89107-282 89107-296 89107-284 89107-298 89107-286 89107-300 89107-288
EcoPolyBlend™ Spill Control Accumulation Centers, Justrite
These handy platforms offer leak protection from drums, pails, batteries, or any other container of hazardous or non-hazardous liquids.
  • Use to create instant spill protection zones and to organize hazardous materials throughout your facility
  • Eco-friendly platforms are made of up to 100% recycled, post-industrial polyethylene
  • All-polyethylene construction provides maximum resistance against chemical attack
  • Multiple centers lock together to hold additional drums
  • Supplied with four removable grates for easy cleanup
High value secondary containment centers are eco-friendly, made of up to 100% recycled, durable polyethylene, black color is 100% recycled; yellow color is 45% recycled. Sturdy, removable grates make sump cleaning easy while low profile offers convenience when loading/unloading. Versatile modular design allows you to customize your workspace around existing structures, linear style, or over entire floor. Optional Joining Clips and Sump-to-Sump™ Drain Kit provide a continuous sump for on-site compliance when needed. Accessory Ramp can be located anywhere on any 2-drum or larger Center.
Certifications: Comply with EPA requirement 40 CFR 264.175, NFPA and SPCC requirements