EcoPolyBlend™ Spill Control Pallets, Justrite®
28631 28624 28626 28622 28628 28623 28635 28637 28636 28633 28630 28634 28627 28632 28629 28635 28625
89107-270EA 591.55 USD
89107-270 89107-250 89107-272 89107-260 89107-252 89107-274 89107-262 89107-276 89107-265 89107-254 89107-275 89107-256 89107-278 89107-266 89107-258 89107-268 89107-248
EcoPolyBlend™ Spill Control Pallets, Justrite®
  • Eco-friendly pallets are made of up to 100% recycled, post-industrial polyethylene
  • UV-protected polyethylene construction provides maximum resistance against chemical attack
  • Self-locating grates remove easily for cleaning
  • Sump catches up to 276L (73gal.) of liquid
  • Comply with EPA requirement 40 CFR 264.175 and the Uniform Fire Code
  • Supplied with drain
Eco-friendly pallets are made of up to 100% recycled polyethylene; dirt hiding black color is 100% recycled, high visibility yellow is 35% recycled content. Fully compliant and quality engineered with a robust design, they withstand the toughest environments and are nearly impervious to chemical attack. Featuring seamless construction, oversized sumps, and sturdy removable self-locating grates, these pallets provide optimum, solid protection from leaks and spills. Two drum and 4-drum square pallets include wide forklift pockets for easy relocation. Optional Ramp for 4-drum square pallets offers convenience when loading and unloading heavy drums. Square or space saving linear styles in multiple sizes offer storage versatility.
Certifications: EPA, SPCC, U, O.