Silicon dioxide ≥99.999% (Si basis), PURATREM, Electronic Grade
Supplier: STREM CHEMICALS INC MS
Synonyms:
Silicon(IV) oxide
, Silicium (IV) oxide
, Silicium dioxide
98-0145-25G
98-0145-100G
98-0145-5G
100201-294EA
489.72
USD
100201-294
100199-214
100205-810
Silicon dioxide ≥99.999% (Si basis), PURATREM, Electronic Grade
Silicium dioxide
Formula:
SiO₂ MW: 60.08 g/mol Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1500 °C Density: 2.6 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 231-545-4 Merck Index: 14,08493 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...