Thorium tetranitrate hydrate ≥99.8% (Th basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Thorium (IV) nitrate tetrahydrate
, Thorium(IV) nitrate hydrate
93-9006-100G
93-9006-25G
100197-976EA
229.29
USD
100197-976
100207-910
Thorium tetranitrate hydrate ≥99.8% (Th basis)
Thorium tetranitrate tetrahydrate
Formula:
N₄O₁₂Th·xH₂O MW: 480.06 g/mol (anhy) |
MDL Number:
MFCD00149432 CAS Number: 13470-07-0 EINECS: 237-514-1 UN: 2910 ADR: 7, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...