Lead titanate ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lead(II) titanate
, Lead titanium oxide
, Lead(II) titanium oxide
, Lead titanate (IV)
, Lead titanate
93-8237-1000G
93-8237-250G
100196-834EA
135.88
USD
100196-834
100200-812
Lead titanate ≥99.5%
Lead titanate
Formula:
O₃PbTi MW: 303.08 g/mol Melting Pt: 1120 °C Density: 7,5 |
MDL Number:
MFCD00011159 CAS Number: 12060-00-3 EINECS: 235-038-9 UN: 3288 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...