Lead(II) bromide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lead dibromide
93-8203-50G
93-8203-250G
100206-062EA
571.82
USD
100206-062
100202-314
Lead(II) bromide ≥98%
Lead (II) bromide
Formula:
Br₂Pb MW: 367.01 g/mol Boiling Pt: 916 °C Melting Pt: 373 °C Density: 6.66 |
MDL Number:
MFCD00011156 CAS Number: 10031-22-8 EINECS: 233-084-4 UN: 2291 ADR: 6.1,III Merck Index: 12,05418 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...