Thallium(I) sulfate ≥99.5% (Tl basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Thallic sulfateThallic sulphateDithallium sulfateDithallium sulphate

93-8121-25G 93-8121-100G
100200-612EA 182.58 USD
100200-612 100200-216
Thallium(I) sulfate ≥99.5% (Tl basis)
Dithallium sulphate
Formula: O₄STl₂
MW: 504.83 g/mol
Boiling Pt: Dec.
Melting Pt: 632 °C
Density: 6.77
Storage Temperature: Ambient
MDL Number: MFCD00011278
CAS Number: 7446-18-6
EINECS: 231-201-3
UN: 1707
ADR: 6.1,II
Merck Index: 12,09405

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR