Thallium(I) fluoride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-8106-10G
93-8106-50G
100202-310EA
336.86
USD
100202-310
100204-438
Thallium(I) fluoride ≥99%
Thallium (I) fluoride
Formula:
FTl MW: 223.38 g/mol Boiling Pt: 655 °C Melting Pt: 327 °C Density: 8.23 |
MDL Number:
MFCD00049607 CAS Number: 7789-27-7 EINECS: 232-154-1 UN: 1707 ADR: 6.1,II Merck Index: 12,09397 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...