Mercury ≥99.998%, redistilled
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-8045-2250G
93-8045-450G
100203-874EA
574.65
USD
100203-874
100201-290
Mercury ≥99.998%, redistilled
Mercury
Formula:
Hg MW: 200.59 g/mol Boiling Pt: 356.9 °C Melting Pt: –38.89 °C Density: 13.546 Storage Temperature: Ambient |
MDL Number:
MFCD00011035 CAS Number: 7439-97-6 EINECS: 231-106-7 UN: 2809 ADR: 8,III Merck Index: 13,05925 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...