Mercury(I) chloride ≥99.5% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Mercurous chloride
, Dimercury dichloride
, Calomel
93-8018-500G
93-8018-100G
100203-872EA
867.63
USD
100203-872
100208-232
Mercury(I) chloride ≥99.5% ACS
Mercury (I) chloride
Formula:
Cl₂Hg₂ MW: 472.09 g/mol Boiling Pt: 384 °C (1013 hPa) Melting Pt: 400 °C (subl.) Density: 7.150 Storage Temperature: Ambient |
MDL Number:
MFCD00011043 CAS Number: 10112-91-1 EINECS: 233-307-5 UN: 2025 ADR: 6.1,III Merck Index: 12,05951 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...