Mercury (II) oxide, red ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-8011-50G
93-8011-250G
100198-056EA
250
USD
100198-056
100210-564
Mercury (II) oxide, red ≥99% ACS
Mercury (II) oxide
Formula:
HgO MW: 216.59 g/mol Boiling Pt: 356.6 °C (1013 hPa) Melting Pt: 500 °C (dec.) Density: 11.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011045 CAS Number: 21908-53-2 EINECS: 244-654-7 UN: 1641 ADR: 6.1,II Merck Index: 13,05908 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...