Gold tribromide, anhydrous ≥99% (99.9+%-Au)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Gold (III) bromide
, Gold(III) bromide
93-7905-5G
93-7905-1G
100201-146EA
174.09
USD
100201-146
100197-002
Gold tribromide, anhydrous ≥99% (99.9+%-Au)
Gold tribromide
Formula:
AuBr₃ MW: 436.68 g/mol Melting Pt: 160 °C |
MDL Number:
MFCD00014171 CAS Number: 10294-28-7 EINECS: 233-654-2 UN: 3260 ADR: 8,II Merck Index: 12,04539 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...