Platinum ≥99.9%, Powder

Supplier: STREM CHEMICALS INC MS

Synonyms: Platinium

93-7830-5G 93-7830-1G
100209-214EA 1321.97 USD
100209-214 100197-960
Platinum ≥99.9%, Powder
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827 °C
Melting Pt: 1769 °C
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR