Dipotassium hexachloroosmate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-7611-250MG
93-7611-1G
93-7611-5G
100206-058EA
1013.42
USD
100206-058
100210-560
100199-188
Dipotassium hexachloroosmate ≥99%
Dipotassium hexachloroosmate
Formula:
Cl₆K₂Os MW: 481.14 g/mol Melting Pt: Dec. Density: 3,42 g/cm³ (20 °C) |
MDL Number:
MFCD00011371 CAS Number: 16871-60-6 EINECS: 240-893-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...