Sodium perrhenate ≥99.9% (Re basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-7508-5G
93-7508-1G
100205-768EA
82.09
USD
100205-768
100200-204
Sodium perrhenate ≥99.9% (Re basis)
Sodium perrhenate
Formula:
NaO₄Re MW: 273.19 g/mol Melting Pt: 300 °C (in O₂) Density: 5.39 |
MDL Number:
MFCD00014231 CAS Number: 13472-33-8 EINECS: 236-742-9 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...