Perrhenic acid ≥99.99% (Re basis) in aqueous solution (50-54% Re), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-7501-10G
93-7501-2G
93-7501-50G
100210-558EA
148.62
USD
100210-558
100209-454
100209-456
Perrhenic acid ≥99.99% (Re basis) in aqueous solution (50-54% Re), PURATREM
Perrhenic acid
Formula:
HO₄Re MW: 251.21 g/mol Melting Pt: 115…125 °C Density: 2.16 g/cm³ (20 °C) |
MDL Number:
MFCD00011326 CAS Number: 13768-11-1 EINECS: 237-380-4 UN: 3264 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...