Tungsten ≥99.95%, wire
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-7452-20MT
93-7452-100MT
100209-212EA
99.08
USD
100209-212
100202-578
Tungsten ≥99.95%, wire
Tungsten
Formula:
W Boiling Pt: 5660 °C Melting Pt: 3410 °C Density: 19,3 |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...