Sodium tungstate dihydrate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Sodium tungsten oxide dihydrateTungstic acid sodium salt dihydrateSodium wolframate dihydrate

93-7422-500G 93-7422-100G
100199-184EA 103.32 USD
100199-184 100200-202
Sodium tungstate dihydrate ≥99% ACS
Sodium wolframate dihydrate
Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 692 °C
Density: 3.24
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
EINECS: 236-743-4
Merck Index: 12,08847

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR