Erbium ≥99.9%, powder
|
Warning
|
93-6830-5G
93-6830-25G
100202-270EA
95
USD
100202-270
100202-720
Erbium ≥99.9%, powder
Erbium
Formula:
Er MW: 167.26 g/mol Boiling Pt: 2863 °C Melting Pt: 1529 °C Density: 9.006 |
MDL Number:
MFCD00010987 CAS Number: 7440-52-0 EINECS: 231-160-1 UN: 3089 ADR: 4,III Merck Index: 12,03679 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...