Erbium trinitrate hydrate ≥99.9% (Er basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Erbium(III) nitrate hydrate
93-6808-25G
93-6808-100G
100204-414EA
84.92
USD
100204-414
100209-450
Erbium trinitrate hydrate ≥99.9% (Er basis)
Erbium trinitrate hydrate
Formula:
Er(NO₃)₃.xH₂O MW: 443.35 g/mol |
MDL Number:
MFCD00149691 CAS Number: 10031-51-3 EINECS: 233-436-7 UN: 1477 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...