Holmium trichloride hexahydrate ≥99.9% (Ho basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6722-2500MG 93-6722-10G 93-6722-50G
100202-262EA 33.97 USD
100202-262 100203-562 100207-306
Holmium trichloride hexahydrate ≥99.9% (Ho basis)
Holmium trichloride hexahydrate
Formula: Cl₃Ho·6H₂O
MW: 379.38 g/mol
MDL Number: MFCD00149739
CAS Number: 14914-84-2
EINECS: 233-387-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR