Dysprosium trifluoride, anhydrous ≥99.9% (dye basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6609-25G
93-6609-100G
100197-262EA
444.43
USD
100197-262
100208-590
Dysprosium trifluoride, anhydrous ≥99.9% (dye basis)
Dysprosium trifluoride
Formula:
DyF₃ MW: 219.5 g/mol Boiling Pt: > 2200 °C Melting Pt: 1360 °C Density: 5.948 g/cm³ (1149 °C) |
MDL Number:
MFCD00016071 CAS Number: 13569-80-7 EINECS: 236-992-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...