Neodymium trinitrate hexahydrate ≥99.9% (Nd basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Neodymium (III) nitrate hexahydrate

93-6008-100G 93-6008-25G
100199-148EA 58.03 USD
100199-148 100208-210
Neodymium trinitrate hexahydrate ≥99.9% (Nd basis)
Neodymium trinitrate hexahydrate
Formula: N₃NdO₉·6H₂O
MW: 438.35 g/mol
Melting Pt: ∼74 °C
Density: 2.265 g/cm³ (20 °C)
MDL Number: MFCD00149799
CAS Number: 16454-60-7
EINECS: 233-153-9
UN: 1477
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR