Neodymium triiodide, anhydrous ≥99.9% (Nd basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6007-5G
93-6007-1G
100204-392EA
290.15
USD
100204-392
100206-160
Neodymium triiodide, anhydrous ≥99.9% (Nd basis)
Neodymium triiodide
Formula:
I₃Nd MW: 524.95 g/mol Melting Pt: 775 °C Density: 5.85 g/cm³ (20 °C) |
MDL Number:
MFCD00016248 CAS Number: 13813-24-6 EINECS: 237-467-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...