Dilanthanum trisulfide ≥99.9% (La basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Lanthanum (III) sulfide
, Lanthanum (III) sulphide
93-5721-25G
93-5721-5G
100204-736EA
162.77
USD
100204-736
100196-770
Dilanthanum trisulfide ≥99.9% (La basis)
Dilanthanum trisulphide
Formula:
La₂S₃ MW: 374.01 g/mol Melting Pt: 2100…2150 °C Density: 4.911 |
MDL Number:
MFCD00016168 CAS Number: 12031-49-1 EINECS: 234-752-8 UN: 3134 ADR: 4.3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...