Lanthanum(III) chloride, anhydrous ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Lanthanum chlorideLanthanum trichloride

93-5708-100G 93-5708-25G
100202-464EA 352 USD
100202-464 100203-864
Lanthanum(III) chloride, anhydrous ≥99.9% (La basis)
Lanthanum (III) chloride
Formula: Cl₃La
MW: 245.26 g/mol
Boiling Pt: 1000 °C
Melting Pt: 860 °C
Density: 3.842
MDL Number: MFCD00011068
CAS Number: 10099-58-8
EINECS: 233-237-5
Merck Index: 13,05379

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR