Barium acetate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Barium di(acetate)Acetic acid barium salt

93-5601-100G 93-5601-500G
100204-732EA 123.14 USD
100204-732 100202-198
Barium acetate ≥99% ACS
Barium acetate
Formula: (H₃CCOO)₂Ba
MW: 255.42 g/mol
Melting Pt: ∼450 °C
Density: 2.468
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,00992

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR