Cesium chloride ≥99.999% (Cs basis), PURATREM
Supplier: STREM CHEMICALS INC MS
Synonyms:
Cesium Chloride
93-5542-10G
93-5542-50G
100209-158EA
174.09
USD
100209-158
100197-900
Cesium chloride ≥99.999% (Cs basis), PURATREM
Cesium chloride
Formula:
CsCl MW: 168.36 g/mol Boiling Pt: 1290 °C Melting Pt: 646 °C Density: 3.988 Storage Temperature: Ambient |
MDL Number:
MFCD00010955 CAS Number: 7647-17-8 EINECS: 231-600-2 Merck Index: 13,02021 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...