Cesium nitrate ≥99.9% (Cs basis)

Warning

Synonyms: Nitric acid cesium salt

93-5527-10G 93-5527-50G
100205-652EA 174.09 USD
100205-652 100204-362
Cesium nitrate ≥99.9% (Cs basis)
Cesium nitrate
Formula: CsNO₃
MW: 194.91 g/mol
Boiling Pt: 737 °C (1013 hPa)
Melting Pt: 414 °C
Density: 3.685
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1,III
Merck Index: 12,02057

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR