Tellurium tetraiodide ≥99.9% (Te basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tellurium(IV) iodide

93-5215-2G 93-5215-10G
100202-192EA 46.71 USD
100202-192 100209-444
Tellurium tetraiodide ≥99.9% (Te basis)
Tellurium tetraiodide
Formula: I₄Te
MW: 635.22 g/mol
Melting Pt: 280 °C
Density: 5.403
MDL Number: MFCD00049578
CAS Number: 7790-48-9
EINECS: 232-210-5
UN: 2923
ADR: 8,II
Merck Index: 12,09280

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR