Antimony(V) chloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Antimony pentachloride
93-5102-100G
93-5102-500G
100206-042EA
288.74
USD
100206-042
100200-772
Antimony(V) chloride ≥99%
Antimony (V) chloride
Formula:
Cl₅Sb MW: 299.02 g/mol Boiling Pt: 79 °C (22 mmHg) Melting Pt: 2.8 °C Density: 2.336 Storage Temperature: Ambient |
MDL Number:
MFCD00011213 CAS Number: 7647-18-9 EINECS: 231-601-8 UN: 1730 ADR: 8,II Merck Index: 13,00701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...