Hexaphenylditin ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-5079-2G 93-5079-10G
100204-730EA 123.14 USD
100204-730 100206-156
Hexaphenylditin ≥99%
Hexaphenylditin
Formula: C₃₆H₃₀Sn₂
MW: 700.05 g/mol
Boiling Pt: 280 °C (dec.)
Melting Pt: 232.5 °C
MDL Number: MFCD00014070
CAS Number: 1064-10-4
EINECS: 213-902-6
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR