Tetraphenyltin ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tetraphenylstannane
93-5052-500G
93-5052-100G
100201-122EA
280.25
USD
100201-122
100207-862
Tetraphenyltin ≥95%
Tetraphenyltin
Formula:
C₂₄H₂₀Sn MW: 427.13 g/mol Boiling Pt: > 420 °C Melting Pt: 226 °C Density: 1.490 |
MDL Number:
MFCD00003006 CAS Number: 595-90-4 EINECS: 209-872-9 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...