Tin(II) chloride, anhydrous ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tin dichloride
, Stannous chloride
93-5030-500G
93-5030-100G
100205-622EA
56.62
USD
100205-622
100199-580
Tin(II) chloride, anhydrous ≥98%
Tin (II) chloride
Formula:
SnCl₂ MW: 189.62 g/mol Boiling Pt: 623 °C Melting Pt: 246.8 °C Density: 3.95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 EINECS: 231-868-0 UN: 3260 ADR: 8,II Merck Index: 12,08939 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...