Indium (III) oxide ≥99.998% (In basis), PURATREM
Supplier: STREM CHEMICALS INC MS
Synonyms:
Indium trioxide
, Diindium trioxide
93-4906-25G
93-4906-5G
100197-240EA
339.69
USD
100197-240
100207-850
Indium (III) oxide ≥99.998% (In basis), PURATREM
Indium (III) oxide
Formula:
In₂O₃ MW: 277.64 g/mol Melting Pt: 715.05 °C Density: 7.179 |
MDL Number:
MFCD00011060 CAS Number: 1312-43-2 EINECS: 215-193-9 Merck Index: 12,04984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...